Learn More
Cayman Chemical Hydroxymethyl Uracil, Purity: ≥98%, CAS: 4433-40-3, 10 g
SDP
Catalog No. 502896620
Encompass_Preferred
Hydroxymethyl uracil is a product of oxidative damage to DNA, predominantly by hydroxyl radical via the Fenton reaction.{2506,2239,2240}
Catalog No. 50-289-6620 Supplier Cayman Chemical Supplier No. 8936010 G
Hydroxymethyl uracil is a product of oxidative damage to DNA, predominantly by hydroxyl radical via the Fenton reaction.{2506,2239,2240}