Learn More
Cayman Chemical Hydroxymethyl UracIl 5g
SDP
Catalog No. 502896331
Encompass_Preferred
Hydroxymethyl uracil is a product of oxidative damage to DNA, predominantly by hydroxyl radical via the Fenton reaction.{2506,2239,2240}
Catalog No. 50-289-6331 Supplier Cayman Chemical Supplier No. 893605 G
Hydroxymethyl uracil is a product of oxidative damage to DNA, predominantly by hydroxyl radical via the Fenton reaction.{2506,2239,2240}