Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Hydroxynaphthol Blue, pH 2 to 3, MilliporeSigma™

Catalog No. 50947643 Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
25 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-947-643 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-947-643 Supplier MilliporeSigma Supplier No. HX08473

missing translation for 'validMainframeMsg'

Specifications

CAS 63451-35-4
Solubility Information 340 g/L
Molecular Weight (g/mol) 620.48
Packaging Glass Bottle
pH 2 to 3 (10 g/L, H2O, 20°C)
Quantity 25 g
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.