Learn More
Medchemexpress LLC Hydroxyphenyl fluorescein | 359010-69-8 | MFCD08062222 | 99.9% | 424.40 | 25 MG
SDP
Catalog No. 5000346858
Encompass_Preferred
Hydroxyphenyl Fluorescein (HPF) is a stable fluorescent probe dye designed for detecting Reactive Oxygen Species (ROS). It offers enhanced specificity and stability compared to H2DCFDA. This probe generates strong green fluorescence when it reacts with intracellular peroxynitroso via a hydroxyl radical reaction. It is suitable for various applications, including fluorescence microscopy, high-throughput imaging, luciferase microplate reader assays, and flow cytometry, with an excitation wavelength of 490 nm and an emission wavelength of 515 nm.
  • Stable ROS fluorescent probe dye.
  • Stronger specificity and stability than H2DCFDA.
  • Produces strong green fluorescence through hydroxyl radical reaction with intracellular peroxynitroso.
  • Applicable for fluorescence microscopy, high-throughput imagers, luciferase microplate readers, or flow cytometry.
  • Excitation/emission of 490/515 nm.
Catalog No. 50-003-46858 Supplier Medchemexpress LLC Supplier No. 25MGHY111330
Hydroxyphenyl Fluorescein (HPF) is a stable fluorescent probe dye designed for detecting Reactive Oxygen Species (ROS). It offers enhanced specificity and stability compared to H2DCFDA. This probe generates strong green fluorescence when it reacts with intracellular peroxynitroso via a hydroxyl radical reaction. It is suitable for various applications, including fluorescence microscopy, high-throughput imaging, luciferase microplate reader assays, and flow cytometry, with an excitation wavelength of 490 nm and an emission wavelength of 515 nm.
  • Stable ROS fluorescent probe dye.
  • Stronger specificity and stability than H2DCFDA.
  • Produces strong green fluorescence through hydroxyl radical reaction with intracellular peroxynitroso.
  • Applicable for fluorescence microscopy, high-throughput imagers, luciferase microplate readers, or flow cytometry.
  • Excitation/emission of 490/515 nm.