Learn More
Medchemexpress LLC Hydroxyphenyl Fluorescein (HPF) | 359010-69-8 | MFCD08062222 | 99.9% | 424.40 | 10 MG
SDP
Catalog No. 5000343367
Encompass_Preferred
Hydroxyphenyl Fluorescein (HPF) is a stable Reactive Oxygen Species (ROS) fluorescent probe dye. It exhibits stronger specificity and stability compared to H2DCFDA. HPF can produce strong green fluorescence through a hydroxyl radical reaction with intracellular peroxynitroso. It can be used for fluorescence microscopy, high-throughput imagers, luciferase microplate readers, or flow cytometry. The excitation/emission wavelengths are 490/515 nm. It is for research use only.
  • Stable ROS fluorescent probe dye.
  • Stronger specificity and stability than H2DCFDA.
  • Produces strong green fluorescence via hydroxyl radical reaction with intracellular peroxynitroso.
  • Applicable for fluorescence microscopy, high-throughput imager, luciferase microplate reader, or flow cytometry.
  • Ex/Em = 490/515 nm.
Catalog No. 50-003-43367 Supplier Medchemexpress LLC Supplier No. HY1113301MG
Hydroxyphenyl Fluorescein (HPF) is a stable Reactive Oxygen Species (ROS) fluorescent probe dye. It exhibits stronger specificity and stability compared to H2DCFDA. HPF can produce strong green fluorescence through a hydroxyl radical reaction with intracellular peroxynitroso. It can be used for fluorescence microscopy, high-throughput imagers, luciferase microplate readers, or flow cytometry. The excitation/emission wavelengths are 490/515 nm. It is for research use only.
  • Stable ROS fluorescent probe dye.
  • Stronger specificity and stability than H2DCFDA.
  • Produces strong green fluorescence via hydroxyl radical reaction with intracellular peroxynitroso.
  • Applicable for fluorescence microscopy, high-throughput imager, luciferase microplate reader, or flow cytometry.
  • Ex/Em = 490/515 nm.