Learn More
Cayman Chemical Hydroxyprogesterone Caproate, Purity: ≥98%, CAS: 630-56-8, 500 mg
SDP
Catalog No. 502935706
Encompass_Preferred
A synthetic progestogen; reduces TNF-α-induced hypertension and decreases renal ET-1 production in pregnant rats (3.32 mg/kg); reduces RUPP-induced hypertension and production of TNF-α and IL-6 in pregnant rats; prevents L-NAME-induced preterm birth in pregnant mice
Catalog No. 50-293-5706 Supplier Cayman Chemical Supplier No. 23978500 MG
A synthetic progestogen; reduces TNF-α-induced hypertension and decreases renal ET-1 production in pregnant rats (3.32 mg/kg); reduces RUPP-induced hypertension and production of TNF-α and IL-6 in pregnant rats; prevents L-NAME-induced preterm birth in pregnant mice