Learn More
Cell Biolabs Inc Hydroxyproline Assay Kit, 96 assays
Catalog No. 501098802
Encompass_Preferred
Hydroxyproline is an amino acid found almost exclusively in collagen protein. Cell Biolabs' Hydroxyproline Assay Kit is a quantitative colorimetric assay for measuring the hydroxyproline concentration in protein samples. The kit detects hydroxyproline concentrations as low as 6.25 µg/mL. Each kit provides sufficient reagents for 96 assays including standards and unknown samples.
Catalog No. 50-109-8802 Supplier Cell Biolabs Inc Supplier No. STA675
Hydroxyproline is an amino acid found almost exclusively in collagen protein. Cell Biolabs' Hydroxyproline Assay Kit is a quantitative colorimetric assay for measuring the hydroxyproline concentration in protein samples. The kit detects hydroxyproline concentrations as low as 6.25 µg/mL. Each kit provides sufficient reagents for 96 assays including standards and unknown samples.