Learn More
Medchemexpress LLC β-Hydroxypropiovanillone | 2196-18-1 | 5 MG
SDP
Catalog No. 5000421670
Encompass_Preferred
β-Hydroxypropiovanillone, a natural compound, shows significant concentration-dependent inhibitory effects on α-glucosidase with an IC50 of 257.8 μg/mL. It is for research use only and not sold to patients.
  • Natural compound
  • Inhibitory effects on α-glucosidase with an IC50 of 257.8 μg/mL
Catalog No. 50-004-21670 Supplier Medchemexpress LLC Supplier No. HYN29295MG
β-Hydroxypropiovanillone, a natural compound, shows significant concentration-dependent inhibitory effects on α-glucosidase with an IC50 of 257.8 μg/mL. It is for research use only and not sold to patients.
  • Natural compound
  • Inhibitory effects on α-glucosidase with an IC50 of 257.8 μg/mL