Learn More
Cyclodextrin Technologies Development HYDROXYPROPYL BCD/PHARM GR 1KG
Supplier:  Cyclodextrin Technologies Development THPBP1KG
Encompass
Beta Cyclodextrin derivative for enhanced solubility >50g/100ml - meets USP/EP standards
Catalog No. NC9686303