Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Hydroxypropyl-beta-cyclodextrin, TCI America™
SDP

Catalog No. H097925G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
25 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
H097925G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. H097925G Supplier TCI America Supplier No. H097925G
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 128446-35-5
Molecular Formula C44H75O36
Molecular Weight (g/mol) 1180.05
MDL Number MFCD00074978

Specifications

Color White
Quantity 25 g
Physical Form Crystalline Powder
Chemical Name or Material Hydroxypropyl-beta-cyclodextrin
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.