Learn More
Selleck Chemical LLC Hydroxypropyl Cellulose S4115-50mg
SDP
Catalog No. 503108900
Encompass_Preferred
Hydroxypropyl cellulose (HPC) is a derivative of cellulose with both water solubility and organic solubility it is used as food additive and sieving matrix for DNA separations
Catalog No. 50-310-8900 Supplier Selleck Chemical LLC Supplier No. S411550MG
Hydroxypropyl cellulose (HPC) is a derivative of cellulose with both water solubility and organic solubility it is used as food additive and sieving matrix for DNA separations