Learn More
Medchemexpress LLC Hydroxypropyl cellulose | 9004-64-2 | 97.0% | C6H7O2(OR1)(OR2)(OR3) | 25 G
SDP
Catalog No. 5000213451
Encompass_Preferred
Hydroxypropyl cellulose is a cellulose ether used as a pharmaceutical excipient and viscosity modifier. It functions as a coating agent, emulsifier, suspending agent, tablet binder, thickener, and viscosifier, and is available in multiple viscosity grades for formulation development and controlled-release applications.
  • Acts as a coating agent and film former.
  • Serves as an emulsifier and suspending agent.
  • Provides thickening and viscosity control across grades.
  • Suitable for tablet binding and controlled-release formulations.
  • Offered in multiple packaging sizes for lab and development use.
  • Typically supplied with high purity for research use.
Catalog No. 50-002-13451 Supplier Medchemexpress LLC Supplier No. HY10055725G
Hydroxypropyl cellulose is a cellulose ether used as a pharmaceutical excipient and viscosity modifier. It functions as a coating agent, emulsifier, suspending agent, tablet binder, thickener, and viscosifier, and is available in multiple viscosity grades for formulation development and controlled-release applications.
  • Acts as a coating agent and film former.
  • Serves as an emulsifier and suspending agent.
  • Provides thickening and viscosity control across grades.
  • Suitable for tablet binding and controlled-release formulations.
  • Offered in multiple packaging sizes for lab and development use.
  • Typically supplied with high purity for research use.