Learn More
Medchemexpress LLC Hydroxypropyl cellulose | 9004-64-2 | 97.0% | Not specified (polymer) | 10 G
SDP
Supplier:  Medchemexpress LLC HY10055710G
Encompass_Preferred
Hydroxypropyl cellulose is a cellulose ether used as a pharmaceutical excipient and surfactant for research applications. It is supplied as a solid and functions as a coating agent, emulsifier, suspending agent, tablet binder, thickener, and viscosifier; storage and purity specifications are provided on the product page.
  • Used as a pharmaceutical excipient and surfactant
  • Functions as coating agent, emulsifier, suspending agent, and tablet binder
  • Provides thickening and viscosity control in formulations
  • Supplied as a solid with stated purity for research use
  • Available in multiple package sizes with documented storage instructions
Catalog No. 50-002-11606