Learn More
Medchemexpress LLC Hydroxypropyl cellulose | 9004-64-2 | 97.0% | Variable (polymer); repeating unit: cellulose, 2-hydroxypropyl ether | 50 G
SDP
Catalog No. 5000211605
Encompass_Preferred
Hydroxypropyl cellulose is a cellulose ether polymer used as a pharmaceutical excipient and formulation aid. It provides film-forming, thickening, and binding properties and is supplied as a white to off-white powder in multiple viscosity grades for use in coatings, suspensions, emulsions, and tablet formulations. The material is intended for research use and is characterized by variable molar mass depending on grade.
  • Provides film-forming and coating properties for oral dosage forms.
  • Acts as an emulsifier and stabilizer in liquid formulations.
  • Improves tablet binding and supports controlled-release matrices.
  • Available in multiple viscosity grades for formulation tuning.
  • White to off-white powder with broad solubility characteristics.
  • Tested purity (NMR) 97.0% per certificate of analysis.
Catalog No. 50-002-11605 Supplier Medchemexpress LLC Supplier No. HY10055750G
Hydroxypropyl cellulose is a cellulose ether polymer used as a pharmaceutical excipient and formulation aid. It provides film-forming, thickening, and binding properties and is supplied as a white to off-white powder in multiple viscosity grades for use in coatings, suspensions, emulsions, and tablet formulations. The material is intended for research use and is characterized by variable molar mass depending on grade.
  • Provides film-forming and coating properties for oral dosage forms.
  • Acts as an emulsifier and stabilizer in liquid formulations.
  • Improves tablet binding and supports controlled-release matrices.
  • Available in multiple viscosity grades for formulation tuning.
  • White to off-white powder with broad solubility characteristics.
  • Tested purity (NMR) 97.0% per certificate of analysis.