Learn More
Medchemexpress LLC Hydroxypropyl starch | 9049-76-7 | MFCD00081709 | 780.8 g/mol | C31H56O22 | 25g
SDP
Catalog No. 5000268652
Encompass_Preferred
Hydroxypropyl starch (2-Hydroxypropyl starch) is a type of modified starch used as a food additive Hydroxypropyl starch is not absorbed intact by the gut but is significantly hydrolyzed by intestinal enzymes and then fermented by intestinal microbiota[1][2]
Catalog No. 50-002-68652 Supplier Medchemexpress LLC Supplier No. HYW71817925G
Hydroxypropyl starch (2-Hydroxypropyl starch) is a type of modified starch used as a food additive Hydroxypropyl starch is not absorbed intact by the gut but is significantly hydrolyzed by intestinal enzymes and then fermented by intestinal microbiota[1][2]