Learn More
Medchemexpress LLC Hydroxypropyl tetrahydropyrantriol | 439685-79-7 | 98.0% | 192.21 g/mol | C8H16O5 | 100 MG
SDP
Catalog No. 5000242150
Encompass_Preferred
Pro-xylane (hydroxypropyl tetrahydropyrantriol) is a 30% aqueous cosmetic active and research reagent that supports the skin extracellular matrix. It stimulates biosynthesis pathways in fibroblasts to enhance structural stability of dermal tissue, improves skin hydration and elasticity, and aids collagen and hyaluronic acid production.
Catalog No. 50-002-42150 Supplier Medchemexpress LLC Supplier No. HY108036100MG
Pro-xylane (hydroxypropyl tetrahydropyrantriol) is a 30% aqueous cosmetic active and research reagent that supports the skin extracellular matrix. It stimulates biosynthesis pathways in fibroblasts to enhance structural stability of dermal tissue, improves skin hydration and elasticity, and aids collagen and hyaluronic acid production.