Learn More
Medchemexpress LLC Hydroxypropyl tetrahydropyrantriol | 439685-79-7 | 98.0% | 192.21 g·mol⁻¹ | C8H16O5 | 250 MG
SDP
Catalog No. 5000242463
Encompass_Preferred
Pro-xylane (30% in water) is an aqueous formulation of hydroxypropyl tetrahydropyrantriol used as a cosmetic active to support skin matrix remodeling. It promotes glycosaminoglycan biosynthesis in fibroblasts, enhances dermal collagen and hyaluronic acid production, and is intended for formulation and research use.
  • Stimulates glycosaminoglycan synthesis in fibroblasts
  • Enhances collagen and hyaluronic acid production in the dermis
  • Improves skin hydration and elasticity
  • Reduces appearance of wrinkles and supports skin firmness
  • Supplied as a 30% aqueous solution with high purity
Catalog No. 50-002-42463 Supplier Medchemexpress LLC Supplier No. HY108036250MG
Pro-xylane (30% in water) is an aqueous formulation of hydroxypropyl tetrahydropyrantriol used as a cosmetic active to support skin matrix remodeling. It promotes glycosaminoglycan biosynthesis in fibroblasts, enhances dermal collagen and hyaluronic acid production, and is intended for formulation and research use.
  • Stimulates glycosaminoglycan synthesis in fibroblasts
  • Enhances collagen and hyaluronic acid production in the dermis
  • Improves skin hydration and elasticity
  • Reduces appearance of wrinkles and supports skin firmness
  • Supplied as a 30% aqueous solution with high purity