Learn More
Selleck Chemical LLC (2-Hydroxypropyl)-b-cyclodextrin (HP-b-CD) S4760-1g
SDP
Catalog No. 503110491
Encompass_Preferred
(2-Hydroxypropyl)- -cyclodextrin (HP- -CD HP- -cyclodextrin Hydroxypropyl betadex Hydroxypropyl- -cyclodextrin) is a well-known sugar used in drug delivery genetic vectors environmental protection and the treatment of Niemann-Pick disease type C1 (NPC1) It is an inhibitor of amyloid- aggregation and widely used drug delivery vehicle to improve stability and bioavailability
Catalog No. 50-311-0491 Supplier Selleck Chemical LLC Supplier No. S47601G
(2-Hydroxypropyl)- -cyclodextrin (HP- -CD HP- -cyclodextrin Hydroxypropyl betadex Hydroxypropyl- -cyclodextrin) is a well-known sugar used in drug delivery genetic vectors environmental protection and the treatment of Niemann-Pick disease type C1 (NPC1) It is an inhibitor of amyloid- aggregation and widely used drug delivery vehicle to improve stability and bioavailability