Learn More
Apexbio Technology LLC Hydroxysafflor yellow A 78281-02-4 20mg
SDP
Catalog No. 5000091659
Encompass_Preferred
Hydroxysafflor yellow A (CAS 78281-02-4) is a chalcone glycoside flavonoid derived from Carthamus tinctorius L It modulates multiple cellular signaling pathways influencing oxidative stress response inflammation regulation and tumor cell apoptosis Hydroxysafflor yellow A exerts its biological activity primarily through autophagy-mediated inhibition of cell proliferation and promotion of apoptosis Laboratory studies indicate antioxidant anti-inflammatory and antitumor activities though specific IC50/EC50 values and experimental models are not provided Based on these pharmacological properties Hydroxysafflor yellow A holds research potential in cardiovascular diseases oxidative damage inflammation-associated diseases and cancer
Catalog No. 50-000-91659 Supplier Apexbio Technology LLC Supplier No. N152620
Hydroxysafflor yellow A (CAS 78281-02-4) is a chalcone glycoside flavonoid derived from Carthamus tinctorius L It modulates multiple cellular signaling pathways influencing oxidative stress response inflammation regulation and tumor cell apoptosis Hydroxysafflor yellow A exerts its biological activity primarily through autophagy-mediated inhibition of cell proliferation and promotion of apoptosis Laboratory studies indicate antioxidant anti-inflammatory and antitumor activities though specific IC50/EC50 values and experimental models are not provided Based on these pharmacological properties Hydroxysafflor yellow A holds research potential in cardiovascular diseases oxidative damage inflammation-associated diseases and cancer