Learn More
Biotium Hydroxystilbamidine, 4% in H2O
Catalog No. 501964785
Encompass_Preferred
Hydroxystilbamidine (also called Fluoro-Gold) has been used extensively as a retrograde tracer for neurons and also a histochemical stain. Fluoro-Gold can undergo retrograde axonal transport, it shows extensive filling of dendrites, and has a high resistance to fading. The 4% solution in deionized H2O is a convenient, ready-to-use form. If desired, the solution can be diluted to a lower concentration. For most applications, the typically dye concentration is 2% to 4%. We also sell hydroxystilbamidine in solid form. Properties: Ex/Em = 361/536 nm; Store at 4°C and protect from light; C18H24N4O7S2; MWt: 473; [223769-64-0].Fluoro-Gold is a trademark of Fluorochrome, LLC.
Catalog No. 50-196-4785 Supplier Biotium Supplier No. 80023
Hydroxystilbamidine (also called Fluoro-Gold) has been used extensively as a retrograde tracer for neurons and also a histochemical stain. Fluoro-Gold can undergo retrograde axonal transport, it shows extensive filling of dendrites, and has a high resistance to fading. The 4% solution in deionized H2O is a convenient, ready-to-use form. If desired, the solution can be diluted to a lower concentration. For most applications, the typically dye concentration is 2% to 4%. We also sell hydroxystilbamidine in solid form. Properties: Ex/Em = 361/536 nm; Store at 4°C and protect from light; C18H24N4O7S2; MWt: 473; [223769-64-0].Fluoro-Gold is a trademark of Fluorochrome, LLC.