Learn More
Cayman Chemical HydroxystIlbamIdInmethns 10mg
SDP
Catalog No. 502934628
Encompass_Preferred
A fluorescent neuronal retrograde tracer and nucleic acid dye; labels the neuronal cell body as well as proximal dendrites; ex/em = 323/620 nm, respectively, when visualized in tissue sections; can be used to label DNA and RNA in fixed or unfixed dead cells; ex/em = 360/450 and 600 nm, respectively, when bound to DNA; ex/em = 360/450 nm, respectively, when bound to RNA
Catalog No. 50-293-4628 Supplier Cayman Chemical Supplier No. 2685810 MG
A fluorescent neuronal retrograde tracer and nucleic acid dye; labels the neuronal cell body as well as proximal dendrites; ex/em = 323/620 nm, respectively, when visualized in tissue sections; can be used to label DNA and RNA in fixed or unfixed dead cells; ex/em = 360/450 and 600 nm, respectively, when bound to DNA; ex/em = 360/450 nm, respectively, when bound to RNA