Learn More
Cayman Chemical Hydroxytacrine (maleate), Purity: ≥98%, CAS: 118909-22-1, 100 mg
SDP
Catalog No. 502966261
Encompass_Preferred
A major active metabolite of tacrine; formed via hydroxylation of tacrine by CYP1A1 and CYP1A2; inhibits AChE (IC50 = 4.8 µM); reverses scopolamine-induced memory impairment in mice without inducing acute toxicity at 33 mg/kg
Catalog No. 50-296-6261 Supplier Cayman Chemical Supplier No. 18508100 MG
A major active metabolite of tacrine; formed via hydroxylation of tacrine by CYP1A1 and CYP1A2; inhibits AChE (IC50 = 4.8 µM); reverses scopolamine-induced memory impairment in mice without inducing acute toxicity at 33 mg/kg