Learn More
Cayman Chemical HydroxythIofentnyld5hyd 100ug
SDP
Catalog No. 502944744
Encompass_Preferred
An internal standard for the quantification of β-hydroxythiofentanyl by GC- or LC-MS; provided as an exempt preparation; intended for research and forensic applications
Catalog No. 50-294-4744 Supplier Cayman Chemical Supplier No. 41302100 UG
An internal standard for the quantification of β-hydroxythiofentanyl by GC- or LC-MS; provided as an exempt preparation; intended for research and forensic applications