Learn More
AdipoGen Hydroxytyrosol
SDP
Supplier:  AdipoGen CDXH0155M100
Encompass_Preferred
Chemical. CAS 10597-60-1. Formula C8H10O3. MW 154.16. Synthetic. Phenolic component of olive oil. Metabolite of oleuropein. Antioxidant that inhibits both 12- and 5-lipoxygenase LO activity. The IC50 values for the inhibition of rat platelet 12-LO and rat neutrophil 5-LO are 4.2 and 13µM, respectively. It does not inhibit, and may actually enhance, COX activity. Protects LDL from both biological and chemical oxidation, suggesting a potential mechanism for the protective effects of olive oil against atherosclerosis. Inhibits the rate of cancer cell proliferation and induces cancer cell apoptosis. Also described to induce growth arrest and apoptosis in human colon carcinoma HT-29 cells through activation of the serine/threonine protein phosphatase 2A. Protects beta-Cells from amylin amyloid-Induced cytotoxicity.
Catalog No. 50-256-0707