Learn More
Selleck Chemical LLC Hydroxytyrosol Acetate S3848-5mg
SDP
Supplier:  Selleck Chemical LLC S38485MG
Encompass_Preferred
Hydroxytyrosol acetate (HTy-Ac) an extra virgin olive oil (EVOO) polyphenol exhibits antioxidant and anti-inflammatory effects
Catalog No. 50-310-9052