Learn More
Medchemexpress LLC Hydroxytyrosol acetate | 69039-02-7 | 100 MG
SDP
Catalog No. 5000418654
Encompass_Preferred
Hydroxytyrosol acetate is a prodrug that converts to Hydroxytyrosol in vivo. Hydroxytyrosol, a phenol found in olive oil, alleviates oxidative stress, improves mitochondrial function, and offers neuroprotective effects.
  • Induces apoptosis in cancer cells via ROS production
  • Shows antibacterial and antiviral effects
  • Used in research for cancer, infection, inflammation, immunology, metabolic, neurological, and cardiovascular diseases
  • Applications include colon cancer, diabetes, Alzheimer's disease, and atherosclerosis
Catalog No. 50-004-18654 Supplier Medchemexpress LLC Supplier No. HYN6043100MG
Hydroxytyrosol acetate is a prodrug that converts to Hydroxytyrosol in vivo. Hydroxytyrosol, a phenol found in olive oil, alleviates oxidative stress, improves mitochondrial function, and offers neuroprotective effects.
  • Induces apoptosis in cancer cells via ROS production
  • Shows antibacterial and antiviral effects
  • Used in research for cancer, infection, inflammation, immunology, metabolic, neurological, and cardiovascular diseases
  • Applications include colon cancer, diabetes, Alzheimer's disease, and atherosclerosis