Learn More
Medchemexpress LLC Hydroxytyrosol acetate | 69039-02-7 | 25 MG
SDP
Catalog No. 5000421633
Encompass_Preferred
Hydroxytyrosol acetate is a prodrug of Hydroxytyrosol, a phenol found in olive oil. It is converted into the active form of Hydroxytyrosol in vivo. This compound can alleviate oxidative stress, improve mitochondrial function, and exert neuroprotective effects. It also induces apoptosis in cancer cells via ROS production and demonstrates antibacterial and antiviral effects.
  • Prodrug of Hydroxytyrosol
  • Alleviates oxidative stress
  • Improves mitochondrial function
  • Exerts neuroprotective effects
  • Induces apoptosis in cancer cells
  • Demonstrates antibacterial and antiviral effects
  • Used for research in various diseases including cancer, infection, inflammation, and neurological conditions
Catalog No. 50-004-21633 Supplier Medchemexpress LLC Supplier No. 25MGHYN6043
Hydroxytyrosol acetate is a prodrug of Hydroxytyrosol, a phenol found in olive oil. It is converted into the active form of Hydroxytyrosol in vivo. This compound can alleviate oxidative stress, improve mitochondrial function, and exert neuroprotective effects. It also induces apoptosis in cancer cells via ROS production and demonstrates antibacterial and antiviral effects.
  • Prodrug of Hydroxytyrosol
  • Alleviates oxidative stress
  • Improves mitochondrial function
  • Exerts neuroprotective effects
  • Induces apoptosis in cancer cells
  • Demonstrates antibacterial and antiviral effects
  • Used for research in various diseases including cancer, infection, inflammation, and neurological conditions