Learn More
Medchemexpress LLC Hydroxytyrosol acetate | 69039-02-7 | 5 MG
SDP
Supplier:  Medchemexpress LLC HYN60435MG
Encompass_Preferred
Hydroxytyrosol acetate is a prodrug of Hydroxytyrosol, a phenol found in olive oil. Converting to its active form in vivo, it exhibits antioxidant, neuroprotective, antibacterial, and antiviral activities. It is studied for applications in cancer, infection, inflammation, and neurological diseases.
  • Converts to active Hydroxytyrosol in vivo.
  • Alleviates oxidative stress and improves mitochondrial function.
  • Provides neuroprotective, antibacterial, and antiviral effects.
  • Induces apoptosis in cancer cells via ROS production.
  • Orally active and blood-brain barrier-permeable.
  • Inhibits *Vibrio* growth by increasing bacterial membrane permeability.
  • Reduces thrombosis and lipid oxidation.
  • Alleviates neuronal apoptosis and inflammatory responses.
  • Improves cognitive function in Alzheimer's disease models.
Catalog No. 50-004-19152