Learn More
Medchemexpress LLC Hydroxytyrosol acetate | 69039-02-7 | MFCD11100257 | 98.1% | 196.20 g/mol | C10H12O4 | 10 MG
SDP
Catalog No. 5000289710
Encompass_Preferred
Hydroxytyrosol acetate is a phenolic research compound used as a prodrug of hydroxytyrosol for preclinical and in vitro studies investigating oxidative stress, apoptosis, neuroprotection, inflammation, and metabolic and cardiovascular disease mechanisms.
  • Prodrug of hydroxytyrosol that converts to the active phenol in biological systems.
  • High purity appropriate for research applications.
  • White to off-white solid with high solubility in DMSO.
  • Store protected from light at refrigerated temperatures; solvent storage options extend stability.
  • Offered in small milligram package sizes for laboratory experiments.
Catalog No. 50-002-89710 Supplier Medchemexpress LLC Supplier No. HYN604310MG
Hydroxytyrosol acetate is a phenolic research compound used as a prodrug of hydroxytyrosol for preclinical and in vitro studies investigating oxidative stress, apoptosis, neuroprotection, inflammation, and metabolic and cardiovascular disease mechanisms.
  • Prodrug of hydroxytyrosol that converts to the active phenol in biological systems.
  • High purity appropriate for research applications.
  • White to off-white solid with high solubility in DMSO.
  • Store protected from light at refrigerated temperatures; solvent storage options extend stability.
  • Offered in small milligram package sizes for laboratory experiments.