Learn More
Medchemexpress LLC Hydroxytyrosol Stan 100mg
SDP
Catalog No. 5000669427
Encompass_Preferred
Hydroxytyrosol (Standard) (DOPET (Standard)) is the analytical standard of Hydroxytyrosol (HY-N0570) Hydroxytyrosol is a phenol found in the olive oil Hydroxytyrosol can alleviate oxidative stress and improve mitochondrial function thereby exerting neuroprotective effects Hydroxytyrosol can induce cancer cells apoptosis via inducing ROS production Hydroxytyrosol shows antibacterial and antiviral effect Hydroxytyrosol can be used for the researches of cancer infection inflammation immunology metabolic neurological and cardiovascular disease such as colon cancer diabetes Alzheimer s Disease and atherosclerosis [1][2][3][4][5][6][7][8]
Catalog No. 50-006-69427 Supplier Medchemexpress LLC Supplier No. 100MGHYN0570R
Hydroxytyrosol (Standard) (DOPET (Standard)) is the analytical standard of Hydroxytyrosol (HY-N0570) Hydroxytyrosol is a phenol found in the olive oil Hydroxytyrosol can alleviate oxidative stress and improve mitochondrial function thereby exerting neuroprotective effects Hydroxytyrosol can induce cancer cells apoptosis via inducing ROS production Hydroxytyrosol shows antibacterial and antiviral effect Hydroxytyrosol can be used for the researches of cancer infection inflammation immunology metabolic neurological and cardiovascular disease such as colon cancer diabetes Alzheimer s Disease and atherosclerosis [1][2][3][4][5][6][7][8]