Learn More
Selleck Chemical LLC Hydroxyurea 10mM/1mL 127-07-1 nci-c04831, nsc32065
SDP
Catalog No. 501361749
Encompass_Preferred
Hydroxyurea (nci-c04831, nsc32065) is an antineoplastic agent that inhibits DNA synthesis through the inhibition of ribonucleoside diphosphate reductase. Hydroxyurea activates apoptosis and autophagy. Hydroxyurea is used to treat HIV infection. *For Research & Development use only.  Product is not intended for drug, household, or human consumption.
Catalog No. 50-136-1749 Supplier Selleck Chemical LLC Supplier No. S189610MM/1ML
Hydroxyurea (nci-c04831, nsc32065) is an antineoplastic agent that inhibits DNA synthesis through the inhibition of ribonucleoside diphosphate reductase. Hydroxyurea activates apoptosis and autophagy. Hydroxyurea is used to treat HIV infection. *For Research & Development use only.  Product is not intended for drug, household, or human consumption.