Learn More
Medchemexpress LLC HY-B0313 5g , Hydroxyurea Hydroxycarbamide CAS:127-07-1 Purity:98%
SDP
Catalog No. 501492195
Encompass_Preferred
Medchemexpress, HY-B0313 5g Hydroxyurea Hydroxycarbamide CAS:127-07-1 Formula:CH4N2O2 Purity:98% Hydroxyurea is a cell apoptosis inducer that inhibit DNA synthesis through inhibition of ribonucleotide reductase. Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-149-2195 Supplier Medchemexpress LLC Supplier No. HYB03135G
Medchemexpress, HY-B0313 5g Hydroxyurea Hydroxycarbamide CAS:127-07-1 Formula:CH4N2O2 Purity:98% Hydroxyurea is a cell apoptosis inducer that inhibit DNA synthesis through inhibition of ribonucleotide reductase. Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.