Learn More
Selleck Chemical LLC Hydroxyurea S1896-200mg
SDP
Catalog No. 503108496
Encompass_Preferred
Hydroxyurea is an antineoplastic agent that inhibits DNA synthesis through the inhibition of ribonucleoside diphosphate reductase Hydroxyurea activates apoptosis and autophagy Hydroxyurea is used to treat HIV infection
Catalog No. 50-310-8496 Supplier Selleck Chemical LLC Supplier No. S1896200MG
Hydroxyurea is an antineoplastic agent that inhibits DNA synthesis through the inhibition of ribonucleoside diphosphate reductase Hydroxyurea activates apoptosis and autophagy Hydroxyurea is used to treat HIV infection