Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Hydroxyvalerenic acid | 1619-16-5 | 5 MG
SDP

Catalog No. 5000418522
Encompass_Preferred

Hydroxyvalerenic acid is a compound isolated from Verbena officinalis. It exhibits low toxicity, with IC50 values of 123 and 165 μM against GLC4 and COLO 320 cells, respectively. This product is intended for research use only.

  • Isolated from plants (*Valeriana officinalis Linn.*)
  • Purity of 98.45%
  • Molecular weight: 250.33
  • Chemical formula: C15H22O3
  • Appearance: solid, white to off-white
  • Storage conditions: 4°C, protected from light; in solvent: -80°C for 6 months, -20°C for 1 month (protected from light)
  • Solubility in ethanol: 2.5 mg/mL (9.99 mM; needs ultrasonic and warming)

Catalog No. 50-004-18522 Supplier Medchemexpress LLC Supplier No. HYN72625MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Hydroxyvalerenic acid is a compound isolated from Verbena officinalis. It exhibits low toxicity, with IC50 values of 123 and 165 μM against GLC4 and COLO 320 cells, respectively. This product is intended for research use only.

  • Isolated from plants (*Valeriana officinalis Linn.*)
  • Purity of 98.45%
  • Molecular weight: 250.33
  • Chemical formula: C15H22O3
  • Appearance: solid, white to off-white
  • Storage conditions: 4°C, protected from light; in solvent: -80°C for 6 months, -20°C for 1 month (protected from light)
  • Solubility in ethanol: 2.5 mg/mL (9.99 mM; needs ultrasonic and warming)

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.