Learn More
Cayman Chemical Hydroxyzine, Purity: ≥98%, CAS: 68-88-2, 100 mg
SDP
Catalog No. 502933245
Encompass_Preferred
A histamine H1 receptor antagonist (Ki = 1.9 nM); binds competitively with the H1 receptor inverse agonist mepyramine in polymorphonuclear leukocytes (IC50 = 80 µM); metabolized to cetirizine in vivo; prevents histamine-induced recruitment of rolling leukocytes in rat mesentery post-capillary venules in situ (10 µM); decreases anxiety-like behavior in mice in the elevated plus maze and light-dark exploration test
Catalog No. 50-293-3245 Supplier Cayman Chemical Supplier No. 24039100 MG
A histamine H1 receptor antagonist (Ki = 1.9 nM); binds competitively with the H1 receptor inverse agonist mepyramine in polymorphonuclear leukocytes (IC50 = 80 µM); metabolized to cetirizine in vivo; prevents histamine-induced recruitment of rolling leukocytes in rat mesentery post-capillary venules in situ (10 µM); decreases anxiety-like behavior in mice in the elevated plus maze and light-dark exploration test