Learn More
Apexbio Technology LLC Hydroxyzine 2HCl, 500mg Cas# 2192-20-3
SDP
Catalog No. 501998627
Encompass_Preferred
Hydroxyzine is a histamine H1-receptor antagonist, inhibits binding of [ 3 H]pyrilamine/[ 3 H]desloratadine to human histamine H1 receptor with IC50 of 10 nM/19 nM. 2192-20-3. MFCD00058200 Hydroxyzine is a histamine H1-receptor antagonist, inhibits binding of [3H]pyrilamine/[3H]desloratadine to human histamine H1 receptor with IC50 of 10 nM/19 nM. There are other sizes available. Please inqury us for quote.
Catalog No. 50-199-8627 Supplier Apexbio Technology LLC Supplier No. B1549500
Hydroxyzine is a histamine H1-receptor antagonist, inhibits binding of [ 3 H]pyrilamine/[ 3 H]desloratadine to human histamine H1 receptor with IC50 of 10 nM/19 nM. 2192-20-3. MFCD00058200 Hydroxyzine is a histamine H1-receptor antagonist, inhibits binding of [3H]pyrilamine/[3H]desloratadine to human histamine H1 receptor with IC50 of 10 nM/19 nM. There are other sizes available. Please inqury us for quote.