Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Hydroxyzine-d4 (dihydrochloride) | 1219805-91-0 | 98.0% | 1 MG
SDP

Catalog No. 5000409430
Encompass_Preferred

Hydroxyzine-d4 (dihydrochloride) is a deuterium-labeled form of Hydroxyzine. This compound acts as a heterocyclic histamine H1-receptor antagonist and possesses anticholinergic, anxiolytic, and analgesic properties.

  • Can be used as a tracer.
  • Serves as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS.
  • Targets the H1 receptor.
  • Molecular weight is 451.85.
  • Chemical formula is C21H25D4Cl3N2O2.
  • Purity (NMR) is 98.0%.
  • Recommended storage for powder is -20°C for 3 years.
  • Recommended storage in solvent is -80°C for 6 months or -20°C for 1 month.

Catalog No. 50-004-09430 Supplier Medchemexpress LLC Supplier No. HYB0548AS11MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Hydroxyzine-d4 (dihydrochloride) is a deuterium-labeled form of Hydroxyzine. This compound acts as a heterocyclic histamine H1-receptor antagonist and possesses anticholinergic, anxiolytic, and analgesic properties.

  • Can be used as a tracer.
  • Serves as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS.
  • Targets the H1 receptor.
  • Molecular weight is 451.85.
  • Chemical formula is C21H25D4Cl3N2O2.
  • Purity (NMR) is 98.0%.
  • Recommended storage for powder is -20°C for 3 years.
  • Recommended storage in solvent is -80°C for 6 months or -20°C for 1 month.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.