Learn More
Medchemexpress LLC Hydroxyzine-d4 dihydrochloride | 1219805-91-0 | 98.0% | 5 MG
SDP
Supplier:  Medchemexpress LLC HYB0548AS15MG
Encompass_Preferred
Hydroxyzine-d4 dihydrochloride is a deuterium labeled form of Hydroxyzine dihydrochloride. This orally active benzodiazepine antihistamine agent functions as a histamine H1-receptor and serotonin antagonist. It possesses anxiolytic effects and is utilized in research for generalized anxiety disorder.
  • Can be used as a tracer.
  • Can be used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS.
  • Stable heavy isotopes are incorporated into drug molecules as tracers for quantitation during drug development.
  • Deuteration can affect pharmacokinetic and metabolic profiles of drugs.
Catalog No. 50-004-08170