Learn More
Medchemexpress LLC Hydroxyzine-d8 | 1189480-47-4 | 99.0% | 1 MG
SDP
Catalog No. 5000408390
Encompass_Preferred
Hydroxyzine-d8 is deuterium labeled Hydroxyzine. It functions as a histamine H1-receptor antagonist. This compound is suitable for use as a tracer and as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS.
  • Used as a tracer
  • Internal standard for quantitative analysis
  • Histamine H1-receptor antagonist
  • Targets H1 receptor
  • Involved in GPCR/G protein, immunology/inflammation, and neuronal signaling pathways
Catalog No. 50-004-08390 Supplier Medchemexpress LLC Supplier No. HYB0548S11MG
Hydroxyzine-d8 is deuterium labeled Hydroxyzine. It functions as a histamine H1-receptor antagonist. This compound is suitable for use as a tracer and as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS.
  • Used as a tracer
  • Internal standard for quantitative analysis
  • Histamine H1-receptor antagonist
  • Targets H1 receptor
  • Involved in GPCR/G protein, immunology/inflammation, and neuronal signaling pathways