Learn More
Medchemexpress LLC Hydroxyzine-d8 | 1189480-47-4 | 99.0% | 5 MG
SDP
Catalog No. 5000409388
Encompass_Preferred
Hydroxyzine-d8 is a deuterium-labeled version of Hydroxyzine, acting as a histamine H1-receptor antagonist. This compound is intended for research use only.
  • Can be used as a tracer
  • Can be used as an internal standard for quantitative analysis by methods such as NMR, GC-MS, or LC-MS
Catalog No. 50-004-09388 Supplier Medchemexpress LLC Supplier No. HYB0548S15MG
Hydroxyzine-d8 is a deuterium-labeled version of Hydroxyzine, acting as a histamine H1-receptor antagonist. This compound is intended for research use only.
  • Can be used as a tracer
  • Can be used as an internal standard for quantitative analysis by methods such as NMR, GC-MS, or LC-MS