Learn More
Medchemexpress LLC HY-B0548A 500mg Medchemexpress, Hydroxyzine (dihydrochloride) CAS:2192-20-3 Purity:>98%
SDP
Catalog No. 502082592
Encompass_Preferred
Medchemexpress, HY-B0548A 500mg Hydroxyzine (dihydrochloride) CAS:2192-20-3 Hydroxyzine dihydrochloride, a benzodiazepine antihistamine agent, acts as a orally active histamine H1-receptor and serotonin antagonist. Hydroxyzine dihydrochloride has anxiolytic effect and can be used forthe research of generalised anxiety disorder. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-208-2592 Supplier Medchemexpress LLC Supplier No. HYB0548A500MG
Medchemexpress, HY-B0548A 500mg Hydroxyzine (dihydrochloride) CAS:2192-20-3 Hydroxyzine dihydrochloride, a benzodiazepine antihistamine agent, acts as a orally active histamine H1-receptor and serotonin antagonist. Hydroxyzine dihydrochloride has anxiolytic effect and can be used forthe research of generalised anxiety disorder. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.