Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Hydroxyzine Pamoate, USP, 97-102%, Spectrum™ Chemical
SDP

Catalog No. 18603368 Shop All Spectrum Chemical Manufacturing Corporation Products
Change view
Click to view available options
Quantity:
25 g
100 g
1 kg
Packaging:
Amber Glass Bottle
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
18-603-368 25 g Amber Glass Bottle
18-603-369 100 g Amber Glass Bottle
18-603-370 1 kg Amber Glass Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 18-603-368 Supplier Spectrum Chemical Manufacturing Corporation Supplier No. H110325GM
Get Up to $500 in Chemicals with Purchase
View all promotions
Add to Cart
Add to Cart

H1103, 10246-75-0, C21H27ClN2O2·C23H16O6

Spectrum™ Chemical Hydroxyzine Pamoate, USP is an antihistamine with anticholinergic (drying) and sedative properties. All Spectrum Chemical USP grade products are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.

Chemical Identifiers

CAS 10246-75-0
Molecular Formula C44H43ClN2O8
Molecular Weight (g/mol) 763.28
InChI Key ASDOKGIIKXGMNB-UHFFFAOYNA-N
IUPAC Name 2-(2-{4-[(4-chlorophenyl)(phenyl)methyl]piperazin-1-yl}ethoxy)ethan-1-ol; 4-[(3-carboxy-2-hydroxynaphthalen-1-yl)methyl]-3-hydroxynaphthalene-2-carboxylic acid
SMILES OCCOCCN1CCN(CC1)C(C1=CC=CC=C1)C1=CC=C(Cl)C=C1.OC(=O)C1=C(O)C(CC2=C3C=CC=CC3=CC(C(O)=O)=C2O)=C2C=CC=CC2=C1

Specifications

CAS Max % 100%
Ignition Residue 0.2%
Packaging Amber Glass Bottle
Quantity 25 g
Percent Purity 97 to 102%
Grade USP
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.