Learn More
Macherey-Nagel MACHEREY-NAGEL HYDRODEX BETA-3P GC Column, Capillary GC Column, Chiral Separation, 25 m
Catalog No. NC3355927
Encompass_Preferred
The MACHEREY-NAGEL HYDRODEX BETA-3P GC Capillary Column is engineered for chiral and enantiomer separations in gas chromatography. Its β-cyclodextrin stationary phase provides superior enantioselectivity, allowing for accurate analysis without derivatization. With a length of 25 m and a 0.25 mm inner diameter, this fused silica column guarantees reliable performance backed by quality testing.
Catalog No. NC3355927 Supplier Macherey-Nagel Supplier No. 723358.25
The MACHEREY-NAGEL HYDRODEX BETA-3P GC Capillary Column is engineered for chiral and enantiomer separations in gas chromatography. Its β-cyclodextrin stationary phase provides superior enantioselectivity, allowing for accurate analysis without derivatization. With a length of 25 m and a 0.25 mm inner diameter, this fused silica column guarantees reliable performance backed by quality testing.