Learn More
MSC HYFLEX INTERCEPT CR SLV
Catalog No. 5000796782
Encompass
The HyFlex 11-281 protective arm sleeves with thumb slot combine cut protection with comfort. Designed with INTERCEPT Cut Resistance Technology, they utilize a mix of natural and synthetic fibers to achieve ANSI/ISEA 105-2024 CUT A4 and EN ISO D protection. Made with HPPE yarn, these sleeves offer cool, breathable all-day comfort and feature a thumb slot for secure placement.
  • Innovative knitwork provides advanced defenses
  • HPPE yarn offers cool, breathable comfort
  • Available in multiple width and length options for a perfect fit
  • Made with 16% recycled content, contributing to a reduced carbon footprint
  • Comply with FDA regulations for safe food handling
  • Certified free of substances harmful to human health via Oeko-Tex certification
Catalog No. 50-007-96782 Supplier MSC Supplier No. 13001680
The HyFlex 11-281 protective arm sleeves with thumb slot combine cut protection with comfort. Designed with INTERCEPT Cut Resistance Technology, they utilize a mix of natural and synthetic fibers to achieve ANSI/ISEA 105-2024 CUT A4 and EN ISO D protection. Made with HPPE yarn, these sleeves offer cool, breathable all-day comfort and feature a thumb slot for secure placement.
  • Innovative knitwork provides advanced defenses
  • HPPE yarn offers cool, breathable comfort
  • Available in multiple width and length options for a perfect fit
  • Made with 16% recycled content, contributing to a reduced carbon footprint
  • Comply with FDA regulations for safe food handling
  • Certified free of substances harmful to human health via Oeko-Tex certification