Learn More
MSC Ansell HyFlex 11-812 Work Gloves, Foam Nitrile-Coated, 18 Gauge, Size 10, 1 Pr.
Catalog No. 5000509056
Encompass
These light-duty, 18-gauge nylon and spandex gloves are engineered for precision handling and enhanced tactile sensitivity. Featuring a specialized tear-away design, the knit construction is developed to deliberately tear upon entanglement, minimizing the risk of severe injury when operating rotating machinery. These gloves incorporate advanced ergonomic technology and are certified skin-friendly for comfort during extended use.
  • Foam nitrile palm coating for improved grip
  • Designed with ZONZ™ Comfort Fit technology for reduced stress and increased dexterity
  • Utilizes ERGOFORM™ technology for a tailored fit
  • Latex-free and silicone-free construction
  • Dermatest™-approved and Oekotex™-certified
  • ANSI/ISEA 105-2024 performance: abrasion level 3, cut level A1, and puncture level 1
  • Length: 200 mm to 250 mm
Catalog No. 50-005-09056 Supplier MSC Supplier No. 10268100
These light-duty, 18-gauge nylon and spandex gloves are engineered for precision handling and enhanced tactile sensitivity. Featuring a specialized tear-away design, the knit construction is developed to deliberately tear upon entanglement, minimizing the risk of severe injury when operating rotating machinery. These gloves incorporate advanced ergonomic technology and are certified skin-friendly for comfort during extended use.
  • Foam nitrile palm coating for improved grip
  • Designed with ZONZ™ Comfort Fit technology for reduced stress and increased dexterity
  • Utilizes ERGOFORM™ technology for a tailored fit
  • Latex-free and silicone-free construction
  • Dermatest™-approved and Oekotex™-certified
  • ANSI/ISEA 105-2024 performance: abrasion level 3, cut level A1, and puncture level 1
  • Length: 200 mm to 250 mm