Learn More
Hygiena Transport-Swab

Specifications
Specifications
| Includes | Pre-filled Letheen broth, 10mL |
| Storage Requirements | 2 to 8°C |
| For Use With (Application) | Ideal sampling device for customers looking for an environmental sample collection device |
| Product Type | Transport Swab |
| Sterility | Sterile |
| Shelf Life | 18 Months |
| Tip Material | Rayon |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.