Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Hygiena Transport-Swab
SDP

Catalog No. HTS4
Change view
Click to view available options
Includes:
Pre-filled Letheen broth, 10mL
Pre-filled Letheen broth, 4mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Includes
HTS4 Pre-filled Letheen broth, 4mL
HTS10 Pre-filled Letheen broth, 10mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. HTS4 Supplier Hygiena Supplier No. HTS4
Add to Cart
Add to Cart

Specifications

Shelf Life 18 Months
Tip Material Rayon
Sterility Sterile
Includes Pre-filled Letheen broth, 4mL
Storage Requirements 2 to 8°C
For Use With (Application) Ideal sampling device for customers looking for an environmental sample collection device
Product Type Transport Swab
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.