Learn More
Grainger Vikan Hygiene Bucket, Polypropylene, 3.17 gal., Purple, 1 Each
Catalog No. 502787715
Encompass_Preferred
This hygiene bucket is designed for food manufacturing, food service, and retail environments where hygiene and safety are critical. Engineered for durability, this purple bucket features a sloped back to facilitate easy mixing and stirring, along with a drip-free spout for precise pouring. It includes a sturdy stainless steel handle and a raised hand grip at the base for improved ergonomics.
  • Capacity of 3.17 gal.
  • Features internal graduation markings in liters and gallons.
  • Constructed from polypropylene and stainless steel (AISI 304).
  • Supports hygienic zoning and 5S programs through color-coding.
  • Autoclavable and resistant to harsh cleaning chemicals.
  • Temperature range of -4°F to 212°F.
  • FDA, Halal, and Kosher compliant.
  • Drainage hole prevents water buildup during storage.
Catalog No. 50-278-7715 Supplier Grainger Supplier No. 18G946
This hygiene bucket is designed for food manufacturing, food service, and retail environments where hygiene and safety are critical. Engineered for durability, this purple bucket features a sloped back to facilitate easy mixing and stirring, along with a drip-free spout for precise pouring. It includes a sturdy stainless steel handle and a raised hand grip at the base for improved ergonomics.
  • Capacity of 3.17 gal.
  • Features internal graduation markings in liters and gallons.
  • Constructed from polypropylene and stainless steel (AISI 304).
  • Supports hygienic zoning and 5S programs through color-coding.
  • Autoclavable and resistant to harsh cleaning chemicals.
  • Temperature range of -4°F to 212°F.
  • FDA, Halal, and Kosher compliant.
  • Drainage hole prevents water buildup during storage.