Learn More
MSC Vikan Hygiene Bucket, 3.17 gal., Orange, Polypropylene/SS, 1 ea
Catalog No. 502869621
Encompass
The Vikan Hygiene Bucket, crafted from polypropylene with a stainless steel grip, is designed for demanding applications, including direct food contact. It meets rigorous international compliance standards for safety and reliability in regulated settings.
  • Constructed from durable polypropylene with an orange color masterbatch
  • Equipped with a stainless steel grip (Grade 1.4301 / AISI 304)
  • Complies with EU food contact regulations: (EC) No 1935/2004, (EC) No 2023/2006, (EU) No 10/2011
  • Raw materials meet US FDA 21 CFR parts 170 to 199 standards
  • Approved for contact with aqueous, acidic, alcoholic, fatty, and dry foods
  • Functions across a wide temperature range from -20 °C to 100 °C
  • Autoclavable at 121 °C for sterilization
Catalog No. 50-286-9621 Supplier MSC Supplier No. 55140750
The Vikan Hygiene Bucket, crafted from polypropylene with a stainless steel grip, is designed for demanding applications, including direct food contact. It meets rigorous international compliance standards for safety and reliability in regulated settings.
  • Constructed from durable polypropylene with an orange color masterbatch
  • Equipped with a stainless steel grip (Grade 1.4301 / AISI 304)
  • Complies with EU food contact regulations: (EC) No 1935/2004, (EC) No 2023/2006, (EU) No 10/2011
  • Raw materials meet US FDA 21 CFR parts 170 to 199 standards
  • Approved for contact with aqueous, acidic, alcoholic, fatty, and dry foods
  • Functions across a wide temperature range from -20 °C to 100 °C
  • Autoclavable at 121 °C for sterilization