Learn More
MSC Vikan Hygiene Bucket, 3.17 Gallon, Polypropylene, Pink
Catalog No. 502838150
Encompass
This Hygiene Bucket is a versatile and durable container designed for various applications, including handling cleaning chemicals and both hot and cold ingredients. Its thoughtful design features a drip-free spout and a raised hand grip for ease of use.
  • Made from polypropylene for durability.
  • Features a drip-free spout for controlled pouring.
  • Equipped with a sturdy stainless steel handle.
  • Calibrated for various measuring units.
  • FDA-compliant materials ensure safety.
  • Suitable for use in temperatures ranging from -4°F to 212°F.
  • Can be sterilized via autoclave up to 249.8°F.
  • Flat side design helps prevent spillage and allows for easy storage.
  • Halal and kosher compliant.
Catalog No. 50-283-8150 Supplier MSC Supplier No. 47171889
This Hygiene Bucket is a versatile and durable container designed for various applications, including handling cleaning chemicals and both hot and cold ingredients. Its thoughtful design features a drip-free spout and a raised hand grip for ease of use.
  • Made from polypropylene for durability.
  • Features a drip-free spout for controlled pouring.
  • Equipped with a sturdy stainless steel handle.
  • Calibrated for various measuring units.
  • FDA-compliant materials ensure safety.
  • Suitable for use in temperatures ranging from -4°F to 212°F.
  • Can be sterilized via autoclave up to 249.8°F.
  • Flat side design helps prevent spillage and allows for easy storage.
  • Halal and kosher compliant.